Merck
  • Impairment of glycoprotein fucosylation in rat hippocampus and the consequences on memory formation.
Impairment of glycoprotein fucosylation in rat hippocampus and the consequences on memory formation.
Pharmacology, biochemistry, and behavior (1986-12-01)
R Jork, G Grecksch, H Matthies
ABSTRACT
The intraventricular injection of 2-deoxy-D-galactose led to a dose- and time-dependent decrease in the fucosylation of hippocampal glycoproteins in rats whereas the incorporation of 3H-N-acetyl-glucosamine was not influenced. This effect is not related to an interference with fucose activating or transferring enzymes but can be abolished by an application of D-galactose. Thus, it seems likely that also in brain tissue a deoxy-galactose induced decrease in the fucosylation is due to a hindering of a glycosidic linkage of fucose to the deoxy-sugar incorporated into glycoprotein chains. As a consequence of an intrahippocampal injection of the deoxy-sugar the retention performance of the animals in a foot-shock motivated brightness discrimination task was considerably impaired. But deoxy-galactose is effective only when administered before and immediately after training whereas either a pre- or a post-training injection did not influence the retention performance of the rats. Thus, an effective metabolic inhibition of the glycoprotein completion by the deoxy-sugar starting at the time of training seems to be crucial to interfere with such morphofunctional alterations in the neuronal network underlying the formation of a memory trace.
Sigma-Aldrich
2-Deoxy-D-galactose, 98%