Merck
Fragment-based ligand design of novel potent inhibitors of tankyrases.
Journal of medicinal chemistry (2013-05-16)
E Andreas Larsson, Anna Jansson, Fui Mee Ng, Siew Wen Then, Resmi Panicker, Boping Liu, Kanda Sangthongpitag, Vishal Pendharkar, Shi Jing Tai, Jeffrey Hill, Chen Dan, Soo Yei Ho, Wei Wen Cheong, Anders Poulsen, Stephanie Blanchard, Grace Ruiting Lin, Jenefer Alam, Thomas H Keller, Pär Nordlund
ABSTRACT
Tankyrases constitute potential drug targets for cancer and myelin-degrading diseases. We have applied a structure- and biophysics-driven fragment-based ligand design strategy to discover a novel family of potent inhibitors for human tankyrases. Biophysical screening based on a thermal shift assay identified highly efficient fragments binding in the nicotinamide-binding site, a local hot spot for fragment binding. Evolution of the fragment hit 4-methyl-1,2-dihydroquinolin-2-one (2) along its 7-vector yields dramatic affinity improvements in the first cycle of expansion. A crystal structure of 7-(2-fluorophenyl)-4-methylquinolin-2(1H)-one (11) reveals that the nonplanar compound extends with its fluorine atom into a pocket, which coincides with a region of the active site where structural differences are seen between tankyrases and other poly(ADP-ribose) polymerase (PARP) family members. A further cycle of optimization yielded compounds with affinities and IC50 values in the low nanomolar range and with good solubility, PARP selectivity, and ligand efficiency.
Sigma-Aldrich
ADP-ribosyltransferase C3 from Clostridium botulinum