Merck
  • Design, fabrication, and preparation of synthetic scaffolds for urologic tissue engineering.
Design, fabrication, and preparation of synthetic scaffolds for urologic tissue engineering.
Methods in molecular biology (Clifton, N.J.) (2013-03-16)
Richard G Payne, Toyin A Knight
ABSTRACT
This chapter describes the fabrication of a polyglycolic acid (PGA)-based scaffold used to tissue engineer a Neo-Urinary Conduit™.
Sigma-Aldrich
Polyglycolic acid
Pricing and availability is not currently available.