Merck
  • Dietary exposure of Antarctic krill to p,p'-DDE: uptake kinetics and toxicological sensitivity in a key polar species.
Dietary exposure of Antarctic krill to p,p'-DDE: uptake kinetics and toxicological sensitivity in a key polar species.
Environmental pollution (Barking, Essex : 1987) (2013-01-29)
Anita H Poulsen, Peter F Landrum, So Kawaguchi, Susan M Bengtson Nash
ABSTRACT
This study evaluated the dietary uptake kinetics and sublethal toxicity of p,p'-dichlorodiphenyl dichloroethylene (p,p'-DDE) in Antarctic krill. The uptake rate constant (characterised by the seawater volume stripped of contaminant sorbed to algae) of 200 ± 0.32 mL g(-1) wet weight h(-1), average absorption efficiency of 86 ± 13% and very low elimination rate constant of 5 × 10(-6) ± 0.0031 h(-1) demonstrate the importance of feeding for p,p'-DDE bioaccumulation in Antarctic krill. Faecal egestion of unabsorbed p,p'-DDE of 8.1 ± 2.7% indicates that this pathway contributes considerably to p,p'-DDE sinking fluxes. A median internal effective concentration (IEC50) of 15 mmol/kg lipid weight for complete immobility indicates baseline toxicity and that Antarctic krill exhibit comparable toxicological sensitivity as temperate species under similar 10 d exposure conditions. These findings support the critical body residue approach and provide insight to the role of Antarctic krill in the biogeochemical cycling of p,p'-DDE in the Southern Ocean.
Sigma-Aldrich
1,1-Dichloro-2,2-bis(4-chlorophenyl)ethene, 99%
Supelco
4,4′-DDE, PESTANAL®, analytical standard