Merck
  • Improved microbial electrocatalysis with osmium polymer modified electrodes.
Improved microbial electrocatalysis with osmium polymer modified electrodes.
Chemical communications (Cambridge, England) (2012-09-13)
Sunil A Patil, Kamrul Hasan, Dónal Leech, Cecilia Hägerhäll, Lo Gorton
ABSTRACT
Using the well-known exoelectrogen Shewanella oneidensis MR-1, an osmium redox polymer modified anode exhibited ca. 4-fold increase in current generation. Additionally, a significant decrease in the start-up time for electrocatalysis was observed. The findings suggest that the inherent extracellular electron transfer capabilities of electrogens coupled with such polymers could enhance electrocatalysis.
Sigma-Aldrich
Osmium, powder, 99.9% trace metals basis