Merck
  • Intraspecific variability of the essential oil of Calamintha nepeta subsp. nepeta from Southern Italy (Apulia).
Intraspecific variability of the essential oil of Calamintha nepeta subsp. nepeta from Southern Italy (Apulia).
Natural product research (2012-06-01)
C Negro, S Notarnicola, L De Bellis, A Miceli
ABSTRACT
The essential oil of 46 spontaneous plants of Calamintha nepeta (L.) Savi subsp. nepeta growing wild in Sud, Italy (Salento, Apulia), were investigated by GC/MS. Fifty-seven components were identified in the oil representing over the 98% of the total oil composition. Four chemotypes were identified: piperitone oxide, piperitenone oxide, piperitone-menthone and pulegone.
Product Description
Supelco
(+)-Pulegone, analytical standard
Sigma-Aldrich
(S)-(−)-Pulegone, 98%
Sigma-Aldrich
(R)-(+)-Pulegone, ≥90%
Sigma-Aldrich
(R)-(+)-Pulegone, 85%, technical grade
Sigma-Aldrich
(R)-(+)-Pulegone, ≥90%