Merck
  • Preparation of rotigotine-loaded microspheres and their combination use with L-DOPA to modify dyskinesias in 6-OHDA-lesioned rats.
Preparation of rotigotine-loaded microspheres and their combination use with L-DOPA to modify dyskinesias in 6-OHDA-lesioned rats.
Pharmaceutical research (2012-05-03)
Aiping Wang, Lexi Wang, Kaoxiang Sun, Wanhui Liu, Chunjie Sha, Youxin Li
ABSTRACT
To prepare rotigotine loaded microspheres (RoMS) to achieve continuous dopaminergic stimulation (CDS) for the treatment of Parkinson's disease (PD) and investigate both the therapeutic benefit and inducibility of AIMs of administration of RoMS combination with L-DOPA in 6-OHDA-leisioned rats. Rotigotine was encapsulated into poly(lactic-co-glycolic acid) (PLGA) microspheres by an oil-in-water emulsion solvent evaporation technique. In vitro characteristics and in vivo pharmacokinetics of RoMS either in rat blood or brain (by microdialysis) were investigated. Contraversive rotations and AIMs were observed to investigate the therapeutic benefit and the propensity to induce dyskinesia of RoMS or RoMS combination with L-DOPA in 6-OHDA-lesioned rats. RoMS displayed continuous-release characteristics of rotigotine in animals and exhibited a steady efficacy lasted for 2 weeks in 6-OHDA-lesioned rats. No significant difference of the therapeutic benefit between the treatment of RoMS and pulsatile L-DOPA combination and mono L-DOPA was found. While the dyskinesia was significantly decreased with the treatment of RoMS and pulsatile L-DOPA combination compared to mono L-DOPA. RoMS could supply an alternative of CDS for the treatment of PD and the study indicates a potential advantage of RoMS for the treatment of mild and advanced PD patient in combination with L-DOPA.
Sigma-Aldrich
Rotigotine hydrochloride, ≥98% (HPLC)