Merck
  • Lipolysis and structure controlled drug release from reversed hexagonal mesophase.
Lipolysis and structure controlled drug release from reversed hexagonal mesophase.
Colloids and surfaces. B, Biointerfaces (2012-02-22)
Nissim Garti, Geut Hoshen, Abraham Aserin
ABSTRACT
The present work investigates a system composed of a ternary reversed hexagonal mesophase (H(II)) loaded with a lipase for modulating drug delivery capabilities of the system. Thermomyces lanuginosa lipase was solubilized into H(II) mesophase for the benefits of continuing lipolysis of the lipids, consequently disordering and decomposing the hexagonal mesophase and thereby enhancing the diffusion of the encapsulated drug. A single transition from the H(II) structure to a random micellar phase was detected during the lipolysis. In the first lipolysis stage the hexagonal system (glycerol monooleate, tricaprylin, and water) preserved its symmetry within ca. 200 min. During this step about 40-60% molar of the lipids were hydrolyzed, and a gradual shrinking of the H(II) cylinders (decrease of 8 Å in lattice parameter) was detected. In the second lipolysis stage, the H(II) mesophase gradually disintegrated (faster rate) and the release of a model drug (sodium diclofenac) was significantly enhanced, which was assumed to be lipolysis rate-controlled. After about 15 h the H(II) mesophase was disintegrated into two dispersed immiscible phases. The release obeyed two-step Higuchi kinetics with two consecutive linear correlations of the drug release.
Product Description
Glycerol mono-oleate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Glyceryl trioctanoate, ≥95% (GC)
Sigma-Aldrich
Glyceryl trioctanoate, technical, ≥90% (GC)