Merck
  • Chiral silver phosphate-catalyzed cycloisomeric kinetic resolution of α-allenic alcohols.
Chiral silver phosphate-catalyzed cycloisomeric kinetic resolution of α-allenic alcohols.
Journal of the American Chemical Society (2012-02-15)
Yan Wang, Kuan Zheng, Ran Hong
ABSTRACT
A kinetic resolution of α-allenic alcohols is realized through chiral silver phosphate-catalyzed cycloisomerization with high stereoselectivity (selectivity factor up to 189) and tolerance of a variety of functional groups. A mechanistic model is proposed to interpret the origin of the high stereoselectivity and broad substrate scope.
Sigma-Aldrich
Silver phosphate, 98%