Merck
  • Modulation of the metabolism of airborne pollutants by glucoraphanin-rich and sulforaphane-rich broccoli sprout beverages in Qidong, China.
Modulation of the metabolism of airborne pollutants by glucoraphanin-rich and sulforaphane-rich broccoli sprout beverages in Qidong, China.
Carcinogenesis (2011-11-03)
Thomas W Kensler, Derek Ng, Steven G Carmella, Menglan Chen, Lisa P Jacobson, Alvaro Muñoz, Patricia A Egner, Jian Guo Chen, Geng Sun Qian, Tao Yang Chen, Jed W Fahey, Paul Talalay, John D Groopman, Jian-Min Yuan, Stephen S Hecht
ABSTRACT
Epidemiological evidence has suggested that consumption of a diet rich in cruciferous vegetables reduces the risk of several types of cancers and chronic degenerative diseases. In particular, broccoli sprouts are a convenient and rich source of the glucosinolate, glucoraphanin, which can release the chemopreventive agent, sulforaphane, an inducer of glutathione S-transferases. Two broccoli sprout-derived beverages, one sulforaphane-rich (SFR) and the other glucoraphanin-rich (GRR), were evaluated for pharmacodynamic action in a crossover clinical trial design. Study participants were recruited from the farming community of He Zuo Township, Qidong, China, previously documented to have a high incidence of hepatocellular carcinoma with concomitant exposures to aflatoxin and more recently characterized with exposures to substantive levels of airborne pollutants. Fifty healthy participants were randomized into two treatment arms. The study protocol was as follows: a 5 days run-in period, a 7 days administration of beverage, a 5 days washout period and a 7 days administration of the opposite beverage. Urinary excretion of the mercapturic acids of acrolein, crotonaldehyde, ethylene oxide and benzene were measured both pre- and postinterventions using liquid chromatography tandem mass spectrometry. Statistically significant increases of 20-50% in the levels of excretion of glutathione-derived conjugates of acrolein, crotonaldehyde and benzene were seen in individuals receiving SFR, GRR or both compared with their preintervention baseline values. No significant differences were seen between the effects of SFR versus GRR. Intervention with broccoli sprouts may enhance detoxication of airborne pollutants and attenuate their associated health risks.
Sigma-Aldrich
Crotonaldehyde, predominantly trans, ≥99%, contains 0.1-0.2% BHT as stabilizer, 1% H2O as stabilizer
Sigma-Aldrich
Crotonaldehyde, mixture of cis and trans, ratio of cis- and trans-isomers (~1:20), ≥99.5% (GC)