Merck
  • Influence of microgel architecture and oil polarity on stabilization of emulsions by stimuli-sensitive core-shell poly(N-isopropylacrylamide-co-methacrylic acid) microgels: Mickering versus Pickering behavior?
Influence of microgel architecture and oil polarity on stabilization of emulsions by stimuli-sensitive core-shell poly(N-isopropylacrylamide-co-methacrylic acid) microgels: Mickering versus Pickering behavior?
Langmuir : the ACS journal of surfaces and colloids (2011-07-09)
Sabrina Schmidt, Tingting Liu, Stephan Rütten, Kim-Ho Phan, Martin Möller, Walter Richtering
ABSTRACT
Charged poly(N-isopropylacrylamide-co-methacrylic acid) [P(NiPAM-co-MAA)] microgels can stabilize thermo- and pH-sensitive emulsions. By placing charged units at different locations in the microgels and comparing the emulsion properties, we demonstrate that their behaviors as emulsion stabilizers are very different from molecular surfactants and rigid Pickering stabilizers. The results show that the stabilization of the emulsions is independent of electrostatic repulsion although the presence and location of charges are relevant. Apparently, the charges facilitate emulsion stabilization via the extent of swelling and deformability of the microgels. The stabilization of these emulsions is linked to the swelling and structure of the microgels at the oil-water interface, which depends not only on the presence of charged moieties and on solvent polarity but also on the microgel (core-shell) morphology. Therefore, the internal soft and porous structure of microgels is important, and these features make microgel-stabilized emulsions characteristically different from classical, rigid-particle-stabilized Pickering emulsions, the stability of which depends on the surface properties of the particles.
Sigma-Aldrich
Poly(N-isopropylacrylamide-co-methacrylic acid), methacrylic acid 5 mol %, average Mn 50,000
Sigma-Aldrich
Poly(N-isopropylacrylamide-co-methacrylic acid), methacrylic acid 10 mol %, Mn 60,000