Merck
  • Isolation and immunomodulatory properties of a flavonoid, casticin from Vitex agnus-castus.
Isolation and immunomodulatory properties of a flavonoid, casticin from Vitex agnus-castus.
Phytotherapy research : PTR (2009-05-15)
M Ahmed Mesaik, Shahnaz Murad, Khalid Mohammed Khan, Rasool Bakhsh Tareen, Aqeel Ahmed, Muhammad Iqbal Choudhary
ABSTRACT
Casticin (1), a flavonoid isolated from the aerial parts of Vitex agnus-castus, was found to be a potent immunomodulatory and cytotoxic compound. The activity was tested in vitro for chemiluminescence, chemotaxis, T-cell proliferation and cytotoxicity. Casticin (1) exhibited a significant inhibitory effect on monocyte oxidative burst in a dose dependent manner. It was found to have a significant suppressive effect on the chemotaxic action at higher concentrations on fMLP (10(-8) m) stimulated neutrophils. It also showed a potent suppressive effect on PHA stimulated T-cell (PMBC).
Product Description
Supelco
Casticin, analytical standard
Sigma-Aldrich
Casticin, from Vitex trifolia, ≥98% (HPLC)
Casticin, primary reference standard