Merck
  • A reassessment of the association between azulene and [60]fullerene. Possible pitfalls in the determination of binding constants through fluorescence spectroscopy.
A reassessment of the association between azulene and [60]fullerene. Possible pitfalls in the determination of binding constants through fluorescence spectroscopy.
Chemical communications (Cambridge, England) (2008-10-03)
Lorenzo Stella, Agostina L Capodilupo, Massimo Bietti
ABSTRACT
We show here that the recently reported surprisingly large association constant (K = 7.6 x 10(4) M(-1)) between azulene and [60]fullerene is due to experimental artifacts, pointing out potential errors in the characterization of association equilibria by fluorescence spectroscopy, and suggesting the best experimental practices.
Sigma-Aldrich
Azulene, 99%
Supelco
Azulene, analytical standard