Merck
A concise approach to the polycyclic scaffold of frondosin D.
The Journal of organic chemistry (2008-09-19)
Kye-Simeon Masters, Bernard L Flynn
ABSTRACT
In a study directed at developing a concise approach to the polycyclic core of frondosin D, a Stille-Heck sequence has been identified that gives direct access to the trimethylbicyclo[5.4.0]undecane ring system common to all frondosins.
Sigma-Aldrich
Undecane, ≥99%
Supelco
Undecane, analytical standard