Merck
  • Subinhibitory concentrations of penicillin increase the mutation rate to optochin resistance in Streptococcus pneumoniae.
Subinhibitory concentrations of penicillin increase the mutation rate to optochin resistance in Streptococcus pneumoniae.
The Journal of antimicrobial chemotherapy (2008-08-08)
Paulo R Cortes, Germán E Piñas, Andrea G Albarracin Orio, José R Echenique
ABSTRACT
The aim of this work was to study the effect of subinhibitory concentrations of penicillin, chloramphenicol and erythromycin on the mutation rate of Streptococcus pneumoniae. The mutation rate to rifampicin and optochin resistance was estimated using fluctuation analysis in three capsulated S. pneumoniae strains, cultured both with and without different subinhibitory antibiotic concentrations. The atpAC and rpoB mutations that conferred optochin and rifampicin resistance, respectively, were identified by DNA sequencing. The exposure to subinhibitory concentrations of penicillin increased the mutation rate (expressed as mutation per cell division) to optochin resistance between 2.1- and 3.1-fold for all three strains studied. In contrast, the rifampicin resistance assay showed no significant variations. To analyse the putative cause of the different responses between the optochin and rifampicin tests, mutations that conferred resistance in both cases were analysed. The difference may be explained by the genetic nature of the atpAC mutations, mostly transversions, which are not efficiently repaired by the HexAB mismatch repair system. We demonstrated that subinhibitory concentrations of penicillin significantly increased the mutation rate of S. pneumoniae, suggesting that exposure to this antibiotic could help this pathogen to acquire mutations that confer resistance to other antibiotics. The optochin test was useful to detect this phenomenon and it should be considered for further mutability analysis in S. pneumoniae.
Product Description
Supelco
Ethylhydrocupreine, analytical standard
Supelco
Ethylhydrocupreine hydrochloride, analytical standard