Merck
  • Au(I)-catalyzed efficient synthesis of functionalized bicyclo[3.2.0]heptanes.
Au(I)-catalyzed efficient synthesis of functionalized bicyclo[3.2.0]heptanes.
Journal of the American Chemical Society (2008-05-10)
Guotao Li, Xiaogen Huang, Liming Zhang
ABSTRACT
An efficient Au(I)-catalyzed synthesis of highly strained and functionalized bicyclo[3.2.0]heptanes is developed. Subsequent couplings with various nucleophiles offer additional structural features/complexity. These one-pot, three-component reactions are proposed to proceed via a key 1,3-dipolar cycloaddition between a Au carbenoid-containing carbonyl ylide and ethyl vinyl ether.
Sigma-Aldrich
Ethyl vinyl ether, purum, ≥98.0% (GC)
Sigma-Aldrich
Ethyl vinyl ether, contains 0.1% KOH as stabilizer, 99%