- Use of Z-amino acid-glyceryl esters in protease catalyzed peptide synthesis.
Use of Z-amino acid-glyceryl esters in protease catalyzed peptide synthesis.
Biomedica biochimica acta (1991-01-01)
J Wiese, H G Gattner, H Zahn
PMID1820068
ABSTRACT
The alpha-glyceryl esters of Z-Gly, Z-Phe and Z-Tyr were synthesized and their use for protease catalyzed peptide synthesis was studied. Three enzymes isolated from crude papain were compared in their catalytic potency. Syntheses with alpha-chymotrypsin were performed in a biphasic system.