Merck
  • 1,3-disubstituted 4-aminopiperidines as useful tools in the optimization of the 2-aminobenzo[a]quinolizine dipeptidyl peptidase IV inhibitors.
1,3-disubstituted 4-aminopiperidines as useful tools in the optimization of the 2-aminobenzo[a]quinolizine dipeptidyl peptidase IV inhibitors.
Bioorganic & medicinal chemistry letters (2007-04-10)
Thomas Lübbers, Markus Böhringer, Luca Gobbi, Michael Hennig, Daniel Hunziker, Bernd Kuhn, Bernd Löffler, Patrizio Mattei, Robert Narquizian, Jens-Uwe Peters, Yves Ruff, Hans Peter Wessel, Pierre Wyss
ABSTRACT
In a search for novel DPP-IV inhibitors, 2-aminobenzo[a]quinolizines were identified as submicromolar HTS hits. Due to the difficult synthetic access to this compound class, 1,3-disubstituted 4-aminopiperidines were used as model compounds for optimization. The developed synthetic methodology and the SAR could be transferred to the 2-aminobenzo[a]quinolizine series, leading to highly active DPP-IV inhibitors.
Sigma-Aldrich
1-Aminopiperidine, 97%