Merck
  • Thermodynamic functions and intra-particle mass transfer kinetics of structural analogues of a template on molecularly imprinted polymers in liquid chromatography.
Thermodynamic functions and intra-particle mass transfer kinetics of structural analogues of a template on molecularly imprinted polymers in liquid chromatography.
Journal of chromatography. A (2005-11-22)
Hyunjung Kim, Georges Guiochon
ABSTRACT
The parameters of the thermodynamics and mass transfer kinetics of the structural analogues (L-enantiomers) of the template were measured on an Fmoc-L-tryptophan (Fmoc-L-Trp) imprinted polymer, at different temperatures. The equilibrium isotherm data and the overloaded band profiles of these compounds were measured at temperatures of 298, 313, 323, and 333 K. The isotherm data were modeled. The thermodynamic functions of the different adsorption sites were derived from the isotherm parameters, using van't Hoff plots. The mass transfer parameters were derived by comparing the experimental peak profiles and profiles calculated using the lumped pore diffusion (POR) model for chromatography. These data show that (1) the strength between the substrate molecules and the MIP increases with increasing number of functional groups on the substrates; (2) enthalpy is the driving force for the affinity of the substrates for the MIP; (3) surface diffusion is the dominant mass transfer mechanism of the substrates through the porous MIP. For those substrate molecules that have the same stereochemistry as the template, the energetic surface heterogeneity needs to be incorporated into the surface diffusion coefficients. Heterogeneous surface diffusivities decrease with increasing affinity of the substrates for the MIP.
Sigma-Aldrich
Fmoc-Gly-OH, ≥98.0% (T)