Merck
  • DFT calculations, structural and spectroscopic studies on the products formed between IBr and N,N'-dimethylbenzoimidazole-2(3H)-thione and -2(3H)-selone.
DFT calculations, structural and spectroscopic studies on the products formed between IBr and N,N'-dimethylbenzoimidazole-2(3H)-thione and -2(3H)-selone.
Dalton transactions (Cambridge, England : 2003) (2005-06-18)
M Carla Aragoni, Massimiliano Arca, Francesco Demartin, Francesco A Devillanova, Alessandra Garau, Francesco Isaia, Vito Lippolis, Gaetano Verani
ABSTRACT
The NBO charge distribution calculated at DFT level on the [LEX](+) species [LE=N,N'-dimethylbenzoimidazole-2(3H)-thione (3) and -2(3H)-selone (4)(Scheme 1); X=I, Br] suggests that the most likely products from the reaction 3 of 4 and with IBr are the 10-X-2 charge-transfer (CT) adduct and the 10-Se-3 "T-shaped" hypervalent adduct featuring a linear Br--Se--I system, respectively. This prediction is confirmed by the synthesis, and X-ray diffraction analysis of 3.IBr (I) and 4.I(0.72)Br(1.28)(II). In particular II, is a 10-Se-3 "T-shaped" hypervalent adduct containing an almost linear X--Se--X system [X--Se--X 179.07(3) degrees, X=I(0.36)/Br(0.64)], which is roughly perpendicular to the average plane of the benzoimidazole moiety. The FT-Raman spectra of I and II agree very well with their structural features. In particular, the complexity of the FT-Raman spectrum of II reflects the disorder in the X-ray crystal structure of this compound.
Sigma-Aldrich
Iodine monobromide solution, 1.0 M in methylene chloride
Sigma-Aldrich
Iodine monobromide, 98%