Merck
  • 2-Haloacrylate reductase, a novel enzyme of the medium chain dehydrogenase/reductase superfamily that catalyzes the reduction of a carbon-carbon double bond of unsaturated organohalogen compounds.
2-Haloacrylate reductase, a novel enzyme of the medium chain dehydrogenase/reductase superfamily that catalyzes the reduction of a carbon-carbon double bond of unsaturated organohalogen compounds.
The Journal of biological chemistry (2005-03-23)
Atsushi Kurata, Tatsuo Kurihara, Harumi Kamachi, Nobuyoshi Esaki
ABSTRACT
A soil bacterium, Burkholderia sp. WS, grows on 2-chloroacrylate as the sole carbon source. To identify the enzymes metabolizing 2-chloroacrylate, we carried out comparative two-dimensional gel electrophoresis of the proteins from 2-chloroacrylate- and lactate-grown bacterial cells. As a result, we found that a protein named CAA43 was inducibly synthesized when the cells were grown on 2-chloroacrylate. The CAA43 gene was cloned and shown to encode a protein of 333 amino acid residues (M(r) 35,788) that shared a significant sequence similarity with NADPH-dependent quinone oxidoreductase from Escherichia coli (38.2% identity). CAA43 was overproduced in E. coli and purified to homogeneity. The purified protein catalyzed the NADPH-dependent reduction of the carbon-carbon double bond of 2-chloroacrylate to produce (S)-2-chloropropionate, which is probably further metabolized to (R)-lactate by (S)-2-haloacid dehalogenase in Burkholderia sp. WS. NADH did not serve as a reductant. Despite the sequence similarity to quinone oxidoreductases, CAA43 did not act on 1,4-benzoquinone and 1,4-naphthoquinone. 2-Chloroacrylate analogs, such as acrylate and methacrylate, were also inert as the substrates. In contrast, 2-bromoacrylate served as the substrate. Thus, we named this novel enzyme 2-haloacrylate reductase. This study revealed a new pathway for the degradation of unsaturated organohalogen compounds. It is also notable that the enzyme is useful for the production of (S)-2-chloropropionate, which is used for the industrial production of aryloxyphenoxypropionic acid herbicides.
Sigma-Aldrich
(S)-(−)-2-Chloropropionic acid, 99%
Sigma-Aldrich
2-Chloropropionic acid, 92%