Merck
  • Isolation and characterization of efficient isoxaben-transforming Microbacterium sp strains from four European soils.
Isolation and characterization of efficient isoxaben-transforming Microbacterium sp strains from four European soils.
Pest management science (2002-12-13)
Sandra Arrault, Stephane Desaint, Colette Catroux, Etienne Sémon, Christian Mougin, Jean Claude Fournier
ABSTRACT
Nutrient-agar plates containing isoxaben (500 mg litre(-1)) were used to isolate isoxaben-metabolising bacteria from four European soils incubated with the herbicide under laboratory conditions. In flask experiments, inoculation of a basal salts medium containing nitrogen and [phenyl-U-14C]isoxaben with an isolate (B2b) resulted in 33% recovery of the initial radioactivity as [14C]carbon dioxide after 2 weeks. A major metabolite identified by GC-MS and NMR analysis as 3-(1-ethyl-1-methylpropyl)isoxazol-5-ylamine accumulated both in basal salts and nutrient broth media. 2,6-Dimethoxybenzoic acid, a suspected metabolite of isoxaben, was not detected in either liquid media. However, the capability of the B2b isolate to use 2,6-dimethoxybenzoic acid as a source of carbon was demonstrated. Soil inoculation with the B2b strain resulted in an increase in the recovery of [14C] carbon dioxide from both [phenyl-U-14C] and [isoxazole-5-14C]isoxaben. The metabolite identified as 3-(1-ethyl-1-methylpropyl)isoxazole-5-ylamine only accumulated if the soil was autoclaved before inoculation. This metabolite was rapidly mineralized by the microflora of a natural soil without history of isoxaben treatment. Homology patterns of sequenced 16S rDNA between isoxaben-transforming isolates and reference strains showed that the four isolates identified belonged to the genus Microbacterium.
Sigma-Aldrich
2,6-Dimethoxybenzoic acid, 98%