- Time-dependent solubilization of IgG in AOT-brine-isooctane microemulsions: role of cluster formation.
Time-dependent solubilization of IgG in AOT-brine-isooctane microemulsions: role of cluster formation.
The stability and structure of protein-containing water-in-oil (w/o) microemulsions were investigated by using the large protein immunoglobulin G (IgG, MW 155,000) in a mixture comprised of brine, sulfosuccinic acid bis [2-ethylhexyl]ester (sodium salt), and isooctane. We explored factors affecting the initial uptake of IgG into the w/o microemulsion and its subsequent release to a solid (precipitate) phase, and the kinetics of the latter process. Influences of such parameters as pH, ionic strength, and protein concentration on the solubilization and precipitation of bovine IgG in the organic phase are described. The structure and dynamics in microemulsions containing bovine IgG were probed by using dynamic light scattering, and it was found that the presence of IgG in the microemulsion induced strong attractive forces between the droplets. Based on results obtained by using these various experimental approaches, a model for protein solubilization and release is proposed. In this model, we propose the formation of clusters within which bovine IgG resides and which substantially slow the kinetics of protein release from the droplets to the precipitate phase.