Merck
  • Biotransformations catalyzed by multimeric enzymes: stabilization of tetrameric ampicillin acylase permits the optimization of ampicillin synthesis under dissociation conditions.
Biotransformations catalyzed by multimeric enzymes: stabilization of tetrameric ampicillin acylase permits the optimization of ampicillin synthesis under dissociation conditions.
Biomacromolecules (2001-12-26)
R Fernández-Lafuente, O Hernández-Jústiz, C Mateo, M Terreni, G Fernández-Lorente, M A Moreno, J Alonso, J L García-López, J M Guisan
ABSTRACT
The importance of the stabilization of the quaternary structure of multimeric enzymes has been illustrated using a model reaction with great industrial relevance: the enzymatic synthesis of ampicillin from 6-amino penicillanic acid (6APA) and phenylglycine methyl ester (PGM) catalyzed by the tetrameric enzyme alpha-amino acid ester hydrolase from Acetobacter turbidans. The stabilization of the multimeric structure of the enzyme was achieved by multi-subunit immobilization of the enzyme followed by its further solid-phase chemical intersubunit cross-linking with polyfunctional macromolecules (dextran-aldehyde). This stabilized derivative has permitted the study of the reaction under conditions where nonstabilized enzyme molecules tended to dissociate (e.g., absence of phosphate ions). Synthetic yields improved from around 65%, under conditions where the nonstabilized derivative was stable, to around 85% in conditions where only the stabilized derivative could be utilized (40% methanol and absence of phosphate ions). When using high concentrations of PGM, a significant worsening of the reaction performance was detected with a significant decrease in the yields (below 55%, using 50 mM 6APA and PGM). This problem has been sorted out by using a fed-batch reaction system. By addition of PGM continuously to the reaction mixture (to maintain the concentration between 0.5 and 3 mM), 95% of 6-APA could be transformed to antibiotic (47.5 mM) by only using a 20% excess of acylating ester.
Sigma-Aldrich
(R)-(−)-2-Phenylglycine methyl ester hydrochloride, ≥95%