Merck
  • Biological spin trapping. II. Toxicity of nitrone spin traps: dose-ranging in the rat.
Biological spin trapping. II. Toxicity of nitrone spin traps: dose-ranging in the rat.
Journal of biochemical and biophysical methods (1995-11-01)
E G Janzen, J L Poyer, C F Schaefer, P E Downs, C M DuBose
ABSTRACT
To obtain the strongest possible free radical spin adduct signal using the electron paramagnetic resonance spectroscopy-spin trapping technique, it is desirable to load an animal with the highest dose of spin trap possible. One hundred and twenty six male Sprague-Dawley rats were used to establish the toxic dose range for PBN (alpha-phenyl N-tert butyl nitrone) and 18 other similar spin traps. The lethal dose of PBN was found to be approximately 100 mg/100 g BW (0.564 mmol/100 g. The 18 other compounds were then tested, and their toxicities were gauged in terms of molar equivalents to PBN. Of these spin traps, DMPO (5,5-dimethyl-1-pyrroline-N-oxide) was found to be the least toxic (no toxic signs at twice the lethal dose for PBN) while 2,6-difluoro-PBN and M4PO (3,3,5,5-tetramethyl-1-pyrroline-N-oxide) were the most toxic, both causing death at one eighth the PBN-equivalent lethal dose. Nine of the 18 nitrones appeared non-toxic at the 0.25 PBN-equivalent lethal dose level.
Sigma-Aldrich
3,3,5,5-Tetramethyl-1-pyrroline N-oxide, 95%