Merck
  • Bacterial consortium proteomics under 4-chlorosalicylate carbon-limiting conditions.
Bacterial consortium proteomics under 4-chlorosalicylate carbon-limiting conditions.
Proteomics (2009-04-22)
Roberto A Bobadilla Fazzini, Agata Bielecka, Ana K Poucas Quintas, Peter N Golyshin, Maria J Preto, Kenneth N Timmis, Vitor A P Martins dos Santos
ABSTRACT
In this study, the stable consortium composed by Pseudomonas reinekei strain MT1 and Achromobacter xylosoxidans strain MT3 (cell numbers in proportion 9:1) was under investigation to reveal bacterial interactions that take place under severe nutrient-limiting conditions. The analysis of steady states in continuous cultures was carried out at the proteome, metabolic profile, and population dynamic levels. Carbon-limiting studies showed a higher metabolic versatility in the community through upregulation of parallel catabolic enzymes (salicylate 5-hydroxylase and 17-fold on 2-keto-4-pentenoate hydratase) indicating a possible alternative carbon routing in the upper degradation pathway highlighting the effect of minor proportions of strain MT3 over the major consortia component strain MT1 with a significant change in the expression levels of the enzymes of the mainly induced biodegradation pathway such as salicylate 1-hydroxylase and catechol 1,2-dioxygenase together with important changes in the outer membrane composition of P. reinekei MT1 under different culture conditions. The study has demonstrated the importance of the outer membrane as a sensing/response protective barrier caused by interspecies interactions highlighting the role of the major outer membrane proteins OprF and porin D in P. reinekei sp. MT1 under the culture conditions tested.
Sigma-Aldrich
4-Chlorosalicylic acid, 93%