Merck
  • Comparison of chromogens for the determination of horseradish peroxidase as a marker in enzyme immunoassay.
Comparison of chromogens for the determination of horseradish peroxidase as a marker in enzyme immunoassay.
Journal of clinical chemistry and clinical biochemistry. Zeitschrift fur klinische Chemie und klinische Biochemie (1981-07-01)
B Porstmann, T Porstmann, E Nugel
ABSTRACT
o-Phenylenediamine, 2,2'-azino-di(3-ethylbenzthiazoline sulphonic acid-6) (ABTS), o-dianisidine and 4-aminoantipyrine were compared as chromogens for the determination of horseradish peroxidase. Highest sensitivity in the determination of horseradish peroxidase-IgG conjugates in dissolved form was obtained with o-phenylenediamine. When these conjugates were used in a two-site binding enzyme immunoassay for hepatitis B surface antigen (HBsAg), the steepest calibration curve and the lowest detection limit were obtained when ABTS was used to determine the immune complexes bound to the solid phase. Non-ionic detergents, such as polyoxyethylene-sorbitol ester, retarded horseradish peroxidase inactivation, resulting in a chromogen-dependent activity rise of horseradish peroxidase. An optimised determination of horseradish peroxidase is reported, in which the sensitivity of the solid phase enzyme immunoassay is doubled by the use of o-dianisidine.
Sigma-Aldrich
2,2′-Azino-bis(3-ethylbenzothiazoline-6-sulfonic acid) diammonium salt, tablet, 10 mg substrate per tablet
Pricing and availability is not currently available.