Merck
  • Prenatal chromosomal microarray analysis in a diagnostic laboratory; experience with >1000 cases and review of the literature.
Prenatal chromosomal microarray analysis in a diagnostic laboratory; experience with >1000 cases and review of the literature.
Prenatal diagnosis (2012-04-03)
Amy Breman, Amber N Pursley, Patricia Hixson, Weimin Bi, Patricia Ward, Carlos A Bacino, Chad Shaw, James R Lupski, Arthur Beaudet, Ankita Patel, Sau W Cheung, Ignatia Van den Veyver
ABSTRACT
To evaluate the results of prenatal chromosomal microarray analysis (CMA) on >1000 fetal samples referred for testing at our institution and to compare these data to published reports. High resolution CMA was offered to women undergoing amniocentesis or chorionic villus sampling. Parental samples were obtained concurrently to exclude maternal cell contamination and assist interpretation of copy number variations. Clinically significant copy number variations were observed in 85/1115 cases (7.6%) overall, and in 45/1075 cases (4.2 %) if 40 abnormal cases with known chromosome abnormalities or familial genomic imbalances were excluded. Eighteen of the 1115 cases had variants of unclear clinical significance (1.6%). Indications yielding the most clinically significant findings were abnormal karyotype/fluorescence in situ hybridization (26/61, 42.6%), family history of chromosomal abnormality (13/137, 9.5%), abnormal ultrasound (38/410, 9.3%), abnormal serum screening (2/37, 5.4%) and advanced maternal age (5/394, 1.3%). Of 1075 cases having no previously known cytogenetic abnormality or family history, 18 (1.7%) had clinically significant genomic changes undetectable by conventional prenatal chromosome analysis. Current experience confirms that the detection rate of CMA for prenatal chromosomal abnormalities surpasses that of conventional karyotype analysis and continues to improve with higher resolution arrays, while maintaining a low frequency of results of unclear clinical significance.