Merck
  • Genetic engineering of the phosphocarrier protein NPr of the Escherichia coli phosphotransferase system selectively improves sugar uptake activity.
Genetic engineering of the phosphocarrier protein NPr of the Escherichia coli phosphotransferase system selectively improves sugar uptake activity.
The Journal of biological chemistry (2012-07-07)
Yossef Lopez-de Los Santos, Henry Chan, Vito A Cantu, Rachael Rettner, Filiberto Sanchez, Zhongge Zhang, Milton H Saier, Xavier Soberon
ABSTRACT
The Escherichia coli phosphoenolpyruvate:sugar phosphotransferase system (PTS) in prokaryotes mediates the uptake and phosphorylation of its numerous substrates through a phosphoryl transfer chain where a phosphoryl transfer protein, HPr, transfers its phosphoryl group to any of several sugar-specific Enzyme IIA proteins in preparation for sugar transport. A phosphoryl transfer protein of the PTS, NPr, homologous to HPr, functions to regulate nitrogen metabolism and shows virtually no enzymatic cross-reactivity with HPr. Here we describe the genetic engineering of a "chimeric" HPr/NPr protein, termed CPr14 because 14 amino acid residues of the interface were replaced. CPr14 shows decreased activity with most PTS permeases relative to HPr, but increases activity with the broad specificity mannose permease. The results lead to the proposal that HPr is not optimal for most PTS permeases but instead represents a compromise with suboptimal activity for most PTS permeases. The evolutionary implications are discussed.
Sigma-Aldrich
D-Mannose 6-phosphate sodium salt, ≥98% (HPLC)