Merck
  • Identification of DDX60 as a Regulator of MHC-I Class Molecules in Colorectal Cancer.
Identification of DDX60 as a Regulator of MHC-I Class Molecules in Colorectal Cancer.
Biomedicines (2022-12-24)
Nina Geng, Tuo Hu, Chunbo He
ABSTRACT
Immune checkpoint blockade (ICB) therapies induce durable responses in approximately 15% of colorectal cancer (CRC) patients who exhibit microsatellite instability-high (MSI-H) or deficient mismatch repair (dMMR). However, more than 80% of CRC patients do not respond to current immunotherapy. The main challenge with these patients is lack of MHC-I signaling to unmask their cancer cells so the immune cells can detect them. Here, we started by comparing IFNγ signature genes and MHC-I correlated gene lists to determine the potential candidates for MHC-I regulators. Then, the protein expression level of listed potential candidates in normal and cancer tissue was compared to select final candidates with enough disparity between the two types of tissues. ISG15 and DDX60 were further tested by wet-lab experiments. Overexpression of DDX60 upregulated the expression of MHC-I, while knockdown of DDX60 reduced the MHC-I expression in CRC cells. Moreover, DDX60 was downregulated in CRC tissues, and lower levels of DDX60 were associated with a poor prognosis. Our data showed that DDX60 could regulate MHC-I expression in CRC; thus, targeting DDX60 may improve the effects of immunotherapy in some patients.