Merck
  • Serological biomarkers of type I, III and IV collagen turnover are associated with the presence and future progression of stricturing and penetrating Crohn's disease.
Serological biomarkers of type I, III and IV collagen turnover are associated with the presence and future progression of stricturing and penetrating Crohn's disease.
Alimentary pharmacology & therapeutics (2022-06-07)
Arno R Bourgonje, Marta S Alexdottir, Antonius T Otten, Roberta Loveikyte, Anne-Christine Bay-Jensen, Martin Pehrsson, Hendrik M van Dullemen, Marijn C Visschedijk, Eleonora A M Festen, Rinse K Weersma, Morten A Karsdal, Klaas Nico Faber, Joachim H Mortensen, Gerard Dijkstra
ABSTRACT
Increased collagen remodelling is a key pathophysiological component underlying intestinal stricture and fistula development in Crohn's disease (CD). To investigate associations between serological biomarkers of collagen turnover and disease behaviour according to the Montreal classification in patients with CD. Serological biomarkers of type III/IV collagen formation (PRO-C3, PRO-C4) and matrix metalloproteinase (MMP) or granzyme-B (GrzB)-mediated type I, III, IV and VI collagen degradation (C1M, C3M, C4M, C4G, C6Ma3) were measured using neo-epitope protein fingerprint assays in 101 patients with CD (Montreal B1: n = 37; B2: n = 27; B3: n = 37) and 96 controls. Patients were followed up until their last outpatient visit to monitor stricturing/penetrating disease progression and recurrence and the occurrence of surgical interventions. C1M, C3M and C4M were significantly reduced in patients with stricturing disease (Montreal B2) and accurately differentiated them from patients with either non-stricturing, non-penetrating (B1) or penetrating (B3) disease (all p < 0.001, multivariable analysis). Similarly, the type IV collagen formation/degradation (PRO-C4/C4M) ratio demonstrated high discriminative capacity (B1/B2: AUC = 0.90; B1/B3: AUC = 0.87, both p < 0.001, multivariable analysis). Prospectively, higher baseline levels of C1M and C4G were associated with an increased risk of penetrating disease progression (C4G: hazard ratio [HR] 1.71 [1.05-2.81], p < 0.05). Elevated degradation of type I, III and IV collagen and excessive (relative) formation of type IV collagen strongly associates with stricturing CD. Type I and IV collagen fragments show predictive potential for the risk of penetrating disease progression. These biomarkers may become valuable tools for detection and prediction of stricturing and penetrating CD.