Merck
  • LncRNA SCIRT is downregulated in atherosclerosis and suppresses the proliferation of human aortic smooth muscle cells (HAOSMCs) by sponging miR-146a in cytoplasm.
LncRNA SCIRT is downregulated in atherosclerosis and suppresses the proliferation of human aortic smooth muscle cells (HAOSMCs) by sponging miR-146a in cytoplasm.
Journal of cardiothoracic surgery (2021-11-08)
Wenhui Gao, Rong Li, Jingjing Yu, Xijie He, Duo Xu, Hai Zhong, Wenwen Dong, Hanbin Cui
ABSTRACT
SCIRT has been characterized as a key player in cancer biology, while its role in other human diseases is unclear. This study explored its role in atherosclerosis, with a specific focus on its interaction with SCIRT and miR-146a. The expression of SCIRT and miR-146a in atherosclerosis-affected tissues and healthy tissues from 56 atherosclerosis patients were analyzed by RT-qPCR. The expression of SCIRT in nuclear and cytoplasm samples was detected by RNA fractionation assay. The direct interaction between SCIRT and miR-146a was detected by RNA pull-down assay. SCIRT and miR-146a were overexpressed in human aortic smooth muscle cells (HAOSMCs) to study the crosstalk between them. The role of SCIRT and miR-146a in the proliferation of HAOSMCs was analyzed with BrdU assay. SCIRT was downregulated by atherosclerosis, while miR-146a was upregulated by atherosclerosis. SCIRT was detected in both cytoplasm and nuclear samples, and it directly interacted with miR-146a. In HAOSMCs, overexpression of SCIRT and miR-146a did not affect the expression of each other. Interestingly, SCIRT suppressed the proliferation of HAOSMCs and reduced the enhancing effects of miR-146a on cell proliferation. Therefore, SCIRT is downregulated in atherosclerosis and it suppresses the proliferation of HAOSMCs by sponging miR-146a in cytoplasm.
Sigma-Aldrich
Human Aortic Smooth Muscle Cells: HAOSMC, adult