Merck
  • PIM447 inhibits oncogenesis and potentiates cisplatin effects in hepatoblastoma.
PIM447 inhibits oncogenesis and potentiates cisplatin effects in hepatoblastoma.
Journal of pediatric surgery (2021-03-26)
Nikita Wadhwani, Hooper R Markert, Raoud Marayati, Laura V Bownes, Colin H Quinn, Jamie M Aye, Jerry E Stewart, Karina J Yoon, Elizabeth A Beierle
ABSTRACT
Novel therapies are needed for patients with hepatoblastoma because of an increasing incidence of disease and poor prognosis for advanced, refractory, and recurrent disease. PIM kinases promote tumorigenesis in hepatoblastoma. A novel PIM inhibitor, PIM447, has shown promise in inhibiting oncogenesis in hematologic and lymphoid malignancies. We hypothesized that PIM inhibition with PIM447 would result in decreased tumorigenesis in hepatoblastoma. The effects of PIM447 on hepatoblastoma viability, proliferation, motility, apoptosis, and tumor cell stemness were assessed in HuH6, a human hepatoblastoma cell line, and COA67, a human hepatoblastoma patient-derived xenograft. PIM447 significantly decreased the viability, proliferation, and motility of HuH6 and COA67 cells. Apoptosis significantly increased following PIM447 treatment. PIM447 had a significant impact on tumor cell stemness as evidenced by decreased expression of CD133 and reduced ability of HuH6 and COA67 cells to form tumorspheres. Furthermore, combining PIM447 with cisplatin resulted in a significant decrease in cell viability compared to either treatment alone. We showed that PIM447 inhibits oncogenesis and potentiates the effects of cisplatin in hepatoblastoma and, therefore, warrants further investigation as a potential therapeutic agent for hepatoblastoma.