Merck
  • Functional CD40 ligand is expressed on human vascular endothelial cells, smooth muscle cells, and macrophages: implications for CD40-CD40 ligand signaling in atherosclerosis.
Functional CD40 ligand is expressed on human vascular endothelial cells, smooth muscle cells, and macrophages: implications for CD40-CD40 ligand signaling in atherosclerosis.
Proceedings of the National Academy of Sciences of the United States of America (1997-03-04)
F Mach, U Schönbeck, G K Sukhova, T Bourcier, J Y Bonnefoy, J S Pober, P Libby
ABSTRACT
Increasing evidence supports involvement of inflammation and immunity in atherogenesis. We report here that CD40 ligand (CD40L), an immunoregulatory signaling molecule heretofore considered largely restricted to recently activated CD4+ T lymphocytes, is expressed by human vascular endothelial cells (EC), smooth muscle cells (SMC), and human macrophages in vitro, and is coexpressed with its receptor CD40 on all three cells types in human atherosclerotic lesions in situ. Cultured human vascular EC, SMC, and human macrophages all constitutively expressed CD40L mRNA as well as protein. Stimulation with interleukin 1beta, tumor necrosis factor alpha, or interferon gamma increased surface levels and de novo synthesis of CD40L on all three cell types. CD40L expressed on EC, SMC, and macrophages exhibited biological activity, as it induced B7.2 expression on B cells. Human vascular SMC also constitutively expressed CD40, the receptor for CD40L, and through CD40 signaling, human recombinant CD40L induced expression of proinflammatory cytokines in these cells, identifying SMC as a target for CD40L. Human atherosclerotic lesions (n = 8) showed expression of immunoreactive CD40L on EC, SMC, and macrophages, while normal arterial tissues (n = 5) contained no CD40L. In atheroma CD40L+ cells often also expressed CD40. These observations establish human vascular EC, SMC, and human macrophages as a novel source of CD40L, and point to T cell-independent CD40 signaling, and a broader function of this pathway in regulation of nonimmune cells, as illustrated here by potential autocrine and paracrine activation during atherogenesis.
Sigma-Aldrich
Monoclonal Anti-CD40 antibody produced in mouse, clone 1G1, purified immunoglobulin, buffered aqueous solution
Pricing and availability is not currently available.