Merck
  • Effects of ractopamine hydrochloride on growth performance, carcass characteristics, and physiological response to different handling techniques.
Effects of ractopamine hydrochloride on growth performance, carcass characteristics, and physiological response to different handling techniques.
Journal of animal science (2017-07-21)
J A Hagenmaier, C D Reinhardt, M J Ritter, M S Calvo-Lorenzo, G J Vogel, C A Guthrie, M G Siemens, K F Lechtenberg, D J Rezac, D U Thomson
ABSTRACT
Feedlot cattle ( = 128; BW = 549 ± 60 kg) were used to evaluate the effects of ractopamine hydrochloride (RAC) on growth performance, physiological response to handling, and mobility during shipment for slaughter in a study utilizing a split-plot design with a 2 × 2 factorial arrangement of treatments: 1) diet (CON [no β-adrenergic agonist] vs. RAC [400 mg·animal·d ractopamine hydrochloride for 28 d]) and 2) handling intensity (HI; low-stress handling [LSH; cattle moved at a walking pace with no electric prod use] vs. high-stress handling [HSH; cattle moved at a minimum of a trot and an electric prod applied while in the alley for posthandling restraint and during loading for shipment to the abattoir]). Cattle fed RAC tended to have greater ADG and G:F ( = 0.06), and had greater HCW and LM area ( = 0.04). The HI treatments were applied on the day after the 28-d growth performance period. Blood samples were collected before HI treatment (baseline), after HI treatments (POSTHAND), after transport to the abattoir (POSTTRANS), and during exsanguination at slaughter. A diet × HI interaction ( = 0.01) was observed in the change in cortisol from baseline to POSTTRANS, and there tended ( ≤ 0.07) to be diet × HI interactions for the change in epinephrine from baseline to POSTHAND and for the change in creatine kinase (CK) from baseline to POSTTRANS. Feeding RAC and HSH both increased the change from baseline to POSTHAND in norepinephrine and pH ( ≤ 0.05). The HSH cattle also had greater changes from baseline to POSTHAND in blood HCO, base excess, partial pressure of CO, lactate, cortisol, and glucose ( ≤ 0.01). Ractopamine and HSH both produced greater increases in CK concentrations from baseline to slaughter ( < 0.01). Mobility was not affected by RAC at the feedlot or following an average 6-h lairage ( ≥ 0.43). This study confirms RAC improves growth performance and suggests metabolic acidosis, a precursor to fatigued cattle syndrome, develops in cattle allowed to trot without the use of a lead rider regardless of RAC administration. Cattle fed RAC displayed altered hormonal responses to handling and transport stress, and the overall proportion of cattle with compromised mobility appears to increase later in the marketing channel. These findings warrant additional research aimed at better understanding the physiological response to stress and protect the welfare of cattle during shipment for slaughter.
Product Description
Supelco
Ractopamine hydrochloride, VETRANAL®, analytical standard