Merck
  • Simultaneous determination of clanobutin, dichlorvos, and naftazone in pork, beef, chicken, milk, and egg using liquid chromatography-tandem mass spectrometry.
Simultaneous determination of clanobutin, dichlorvos, and naftazone in pork, beef, chicken, milk, and egg using liquid chromatography-tandem mass spectrometry.
Food chemistry (2018-02-27)
Jin-A Park, A M Abd El-Aty, Weijia Zheng, Seong-Kwan Kim, Sang-Hyun Cho, Jeong-Min Choi, Ahmet Hacımüftüo, Ji Hoon Jeong, Jing Wang, Jae-Han Shim, Ho-Chul Shin
ABSTRACT
A chromatographic method involving a single run was validated for the quantification of clanobutin, dichlorvos, and naftazone in products of animal origin. Pork, beef, chicken, milk, and egg samples were extracted with a solution of 0.1% formic acid in acetonitrile, defatted with n-hexane, centrifuged, and filtered prior to analysis using liquid chromatography-tandem mass spectrometry (LC-/MS/MS). The analytes were separated on a C18 column using a solution of 0.1% formic acid and 10 mM ammonium formate (A) and acetonitrile (B) as the mobile phase. A good linearity over 5-50 ng/g concentration range was obtained with coefficients of determination (R2) ≥ 0.9807. The intra- and interday accuracy (recovery %) calculated from 3 fortification levels (5, 10, and 20 ng/g) were 73.2-108.1% and 71.4-109.8%, and the precisions (expressed relative standard deviations (RSDs)) were 0.9-12.9% and 1.8-10.6%, respectively, for the 3 tested analytes in animal originated foods. The limits of quantification (LOQs) ranged between 0.1 and 1 ng/g, thus enabling the quantification of residual levels below the uniform maximum residue limit (MRL) of 0.01 mg/kg set for compounds having no MRL. The designated methodology was successfully applied to monitor various samples collected from Seoul; the tested analytes were not quantified in any of the market samples. Conclusively, the developed method is simple, sensitive, and accurate, and could be used for the detection of pharmaceuticals in various animal food matrices with variable protein and fat contents.
Product Description
Supelco
Trichlorfon, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland