Merck
  • Racemic resolution of some DL-amino acids using Aspergillus fumigatus L-amino acid oxidase.
Racemic resolution of some DL-amino acids using Aspergillus fumigatus L-amino acid oxidase.
Current microbiology (2011-05-19)
Susmita Singh, Binod K Gogoi, Rajib L Bezbaruah
ABSTRACT
The ability of Aspergillus fumigatus L-amino acid oxidase (L-aao) to cause the resolution of racemic mixtures of DL-amino acids was investigated with DL-alanine, DL-phenylalanine, DL-tyrosine, and DL-aspartic acid. A chiral column, Crownpak CR+ was used for the analysis of the amino acids. The enzyme was able to cause the resolution of the three DL-amino acids resulting in the production of optically pure D-alanine (100% resolution), D-phenylalanine (80.2%), and D-tyrosine (84.1%), respectively. The optically pure D-amino acids have many uses and thus can be exploited industrially. This is the first report of the use of A. fumigatus L: -amino acid oxidase for racemic resolution of DL-amino acids.
Sigma-Aldrich
DL-Alanine, ≥99% (HPLC)
Sigma-Aldrich
DL-Aspartic acid, ≥99% (TLC)
Sigma-Aldrich
DL-Aspartic acid potassium salt