Merck
  • Addition of methyl cellulose enema to double-contrast barium imaging of sigmoid diverticulosis.
Addition of methyl cellulose enema to double-contrast barium imaging of sigmoid diverticulosis.
Acta radiologica (Stockholm, Sweden : 1987) (1997-01-01)
R Olsson, I Adnerhill, P Björkdahl, O Ekberg, F T Fork
ABSTRACT
Double-contrast barium enema has a reduced sensitivity in patients with severe sigmoid diverticulosis. Therefore a carboxy methyl cellulose enema was employed after the conventional double-contrast examination in 15 patients with sigmoid diverticulosis. A significant increase in lumen diameter and a superior removal of barium residue from the diverticula facilitated the interpretation of the sigmoid loops. The addition of methyl cellulose enema to double-contrast barium imaging improves diagnostic imaging in diverticulosis by expanding the lumen and emptying the diverticula.
Sigma-Aldrich
Methyl cellulose, viscosity 3000-5500 mPa.s, 2 % in H2O(20 °C)