Merck
  • Fish Oil Diet during Pre-mating, Gestation, and Lactation in Adult Offspring Rats on Cancer Cachexia Prevention.
Fish Oil Diet during Pre-mating, Gestation, and Lactation in Adult Offspring Rats on Cancer Cachexia Prevention.
Molecular nutrition & food research (2021-03-03)
Sarah Christine Pereira de Oliveira, Natália Angelo da Silva Miyaguti, Steven Thomas Russell, Natália Tobar, Murilo Vieira Geraldo, Maria Cristina Cintra Gomes-Marcondes
ABSTRACT
Nutritional supplementation of the maternal diet can modify the cancer susceptibility in adult offspring. Therefore, the authors evaluate the effects of a fish-oil diet administered to a long-term, during pre-mating, gestation, and lactation, in reducing cancer-cachexia damages in adult Walker-256 tumor-bearing offspring. Female rats receive control or fish oil diet during pre-mating, gestation, and lactation. After weaning, male offspring are fed the control diet until adulthood and distributed in (C) control adult-offspring; (W) adult tumor-bearing offspring; (OC) adult-offspring of maternal fish oil diet; (WOC) adult tumor-bearing offspring of maternal fish oil diet groups. Fat body mass is preserved, muscle expression of mechanistic target of rapamicin (mTOR) and eukariotic binding protein of eukariotic factor 4E (4E-BP1) is modified, being associated with lower 20S proteasome protein expression, and the liver alanine aminotransferase (ALT) enzyme content maintained in the WOC group. Also, the OC group shows reduced triglyceridemia. In this experimental model of cachexia, the long-term maternal supplementation is a positive strategy to improve liver function and lipid metabolism, as well as to modify muscle proteins expression in the mTOR pathway and also reduce the 20S muscle proteasome protein, without altering the tumor development and muscle wasting in adult tumor-bearing offspring.