Merck
  • Dose-Dependent Increases in Ellagitannin Metabolites as Biomarkers of Intake in Humans Consuming Standardized Black Raspberry Food Products Designed for Clinical Trials.
Dose-Dependent Increases in Ellagitannin Metabolites as Biomarkers of Intake in Humans Consuming Standardized Black Raspberry Food Products Designed for Clinical Trials.
Molecular nutrition & food research (2020-03-01)
Kristen M Roberts, Elizabeth M Grainger, Jennifer M Thomas-Ahner, Alice Hinton, Junnan Gu, Ken Riedl, Yael Vodovotz, Ronney Abaza, Steven J Schwartz, Steven K Clinton
ABSTRACT
Black raspberry (BRB) phytochemicals demonstrate anti-carcinogenic properties in experimental models, including prostate cancer. Two BRB foods, a confection and nectar, providing a consistent and reproducible product for human clinical studies are designed and characterized. Men with clinically localized prostate cancer are sequentially enrolled to a control group or one of four intervention groups (confection or nectar, 10 or 20 g dose; n = 8 per group) for 4 weeks prior to prostatectomy. Primary outcomes include: safety, adherence, and ellagitannin metabolism. Adherence to the intervention is >96%. No significant (≥grade II) toxicities are detected. Urinary urolithins (A, B, C, and D) and dimethyl ellagic acid (DMEA) quantified by Ultra high performance liquid chromatography tandem mass spectroscopy (UPLC/MS/MS) indicate a dose-dependent excretion yet heterogeneous patterns among men. Men in the BRB confection groups have greater urinary excretion of the microbial urinary metabolites urolithin A and DMEA, suggesting that this food matrix provides greater colonic microflora exposure. Fully characterized BRB confections and nectar are ideal for food-based large phase III human clinical studies. BRB products provide a bioavailable source of BRB phytochemicals, however large inter individual variation in polyphenol metabolism suggests that host genetics, microflora, and other factors are critical to understanding bioactivity and metabolism.