Merck
  • Synthesis of Thermoplastic Poly(2-methoxyethyl acrylate)-Based Polyurethane by RAFT and Condensation Polymerization.
Synthesis of Thermoplastic Poly(2-methoxyethyl acrylate)-Based Polyurethane by RAFT and Condensation Polymerization.
Macromolecular rapid communications (2020-08-19)
Shunsuke Tazawa, Tomoki Maeda, Masamitsu Nakayama, Atsushi Hotta
ABSTRACT
Thermoplastic solid poly(2-methoxyethyl acrylate) (PMEA)-based polyurethane (PU) is synthesized through the reversible addition-fragmentation chain transfer (RAFT) polymerization and the condensation polymerization, using hydroxyl-terminated RAFT reagents and diisocyanate, respectively. Neat PMEA is a promising antithrombogenic liquid used in the medical fields. The thermoplastic property of the solid PMEA-based PU due to hydrogen bonding is confirmed by the dynamic mechanical analysis (DMA) at temperature below 72 °C. The antithrombogenic property of PMEA-based PU is also analyzed by the platelet adhesion test. The number of platelets on PMEA-based PU is 17 cells per unit area, which is smaller than that on the fluorinated diamond-like carbon (F-DLC), a well-known highly antithrombogenic material. It is concluded that a newly synthesized PMEA-based PU exhibits thermoplastic characteristics with excellent antithrombogenicity.
Sigma-Aldrich
2-Nitro-5-(2-propynyloxy)benzyl 4-cyano-4-(phenylcarbonothioylthio)pentanoate, 97%
Sigma-Aldrich
Ethylene glycol methyl ether acrylate, contains 50-100 ppm MEHQ as inhibitor, 98%