Merck
  • Preparation of Functionalized Carbon-Coated Cobalt Nanoparticles with Sulfonated Arene Derivatives, a Study on Surface Functionalization and Stability.
Preparation of Functionalized Carbon-Coated Cobalt Nanoparticles with Sulfonated Arene Derivatives, a Study on Surface Functionalization and Stability.
Chemistry (Weinheim an der Bergstrasse, Germany) (2020-12-23)
Simon Doswald, Wendelin J Stark
ABSTRACT
The functionalization of magnetic nanoparticles has been an important field in the last decade due to the versatile applications in catalysis and biomedicine. Generally, a high degree of functionalities on the surface of the nanoparticles is desired. In this study, covalent functionalization of various aromatic sulfonic acids on carbon-coated cobalt nanoparticles are investigated on surface functionalization yield and stability. The nanoparticles are prepared via covalent linkage of an in situ generated diazonium on the graphene-like surface. Adsorption and wash experiments were performed to confirm a covalent bonding of the naphthalene derivatives on the nanoparticle surface. With an increased number of sulfonic acid groups on the aromatic compound a significantly lower loading is observed on the corresponding functionalized nanoparticles. This can be counteracted by a change of nitrite species. With this method, nanoparticles with a high number of sulfonic acid groups can be produced.
Sigma-Aldrich
3-Aminobenzenesulfonic acid, 97%