Merck
  • Covalent functionalization of molybdenum disulfide by chemically activated diazonium salts.
Covalent functionalization of molybdenum disulfide by chemically activated diazonium salts.
Nanoscale (2021-01-29)
Lakshya Daukiya, Joan Teyssandier, Samuel Eyley, Salim El Kazzi, Miriam Candelaria Rodríguez González, Bapi Pradhan, Wim Thielemans, Johan Hofkens, Steven De Feyter
ABSTRACT
Covalent functionalization is one of the most efficient ways to tune the properties of layered materials in a highly controlled manner. However, molecular chemisorption on semiconducting transition metal dichalcogenides remains a delicate task due to the inertness of their surface. Here we perform covalent modification of bulk and single layer molybdenum disulfide (MoS2) using chemical activation of diazonium salts. A high level of control over the grafting density and yield on MoS2 basal plane can be achieved by this approach. Using scanning probe microscopies and X-ray photoelectron spectroscopy we prove the covalent functionalization of MoS2.
Sigma-Aldrich
3,5-Di-tert-butylaniline, 98%