Merck
  • Portable Real-Time Detection of Pb(II) Using a CMOS MEMS-Based Nanomechanical Sensing Array Modified with PEDOT:PSS.
Portable Real-Time Detection of Pb(II) Using a CMOS MEMS-Based Nanomechanical Sensing Array Modified with PEDOT:PSS.
Nanomaterials (Basel, Switzerland) (2020-12-12)
Yi-Kuang Yen, Chao-Yu Lai
ABSTRACT
Detecting the concentration of Pb2+ ions is important for monitoring the quality of water due to it can become a health threat as being in certain level. In this study, we report a nanomechanical Pb2+ sensor by employing the complementary metal-oxide-semiconductor microelectromechanical system (CMOS MEMS)-based piezoresistive microcantilevers coated with PEDOT:PSS sensing layers. Upon reaction with Pb2+, the PEDOT:PSS layer was oxidized which induced the surface stress change resulted in a subsequent bending of the microcantilever with the signal response of relative resistance change. This sensing platform has the advantages of being mass-produced, miniaturized, and portable. The sensor exhibited its sensitivity to Pb2+ concentrations in a linear range of 0.01-1000 ppm, and the limit of detection was 5 ppb. Moreover, the sensor showed the specificity to Pb2+, required a small sample volume and was easy to operate. Therefore, the proposed analytical method described here may be a sensitive, cost-effective and portable sensing tool for on-site water quality measurement and pollution detection.
Sigma-Aldrich
Poly(3,4-ethylenedioxythiophene)-poly(styrenesulfonate), 1.3 wt % dispersion in H2O
Pricing and availability is not currently available.