Merck
  • Paraoxonase 1 Phenotype and Protein N-Homocysteinylation in Patients with Rheumatoid Arthritis: Implications for Cardiovascular Disease.
Paraoxonase 1 Phenotype and Protein N-Homocysteinylation in Patients with Rheumatoid Arthritis: Implications for Cardiovascular Disease.
Antioxidants (Basel, Switzerland) (2020-09-25)
Jolanta Parada-Turska, Grażyna Wójcicka, Jerzy Beltowski
ABSTRACT
Paraoxonase 1 (PON1) is the high density lipoprotein-associated esterase which inhibits the development of atherosclerosis by metabolizing lipid peroxidation products as well as hydrolyzing proatherogenic metabolite of homocysteine (Hcy), Hcy thiolactone, which otherwise reacts with lysine groups of proteins, thus forming N-Hcy-protein in a process referred to as protein N-homocysteinylation. Rheumatoid arthritis (RA) is the chronic inflammatory autoimmune disease associated with increased risk of cardiovascular complications, but the underlying mechanisms are incompletely understood. We examined PON1 status and N-homocysteinylation of serum proteins in patients with RA. Blood was collected from 74 RA patients and 70 control subjects. PON1 activity was measured toward synthetic (paraoxon, phenyl acetate) and natural (Hcy thiolactone) substrates. PON1 protein concentration was measured by ELISA. Total Hcy as well as N-Hcy-protein were measured in serum as well. PON1 activity toward Hcy thiolactone was lower in RA patients than in control subjects which was accompanied by increased concentration of N-Hcy-protein despite normal total Hcy concentration. PON1 protein concentration was unchanged in the RA group, but the specific enzyme activity was reduced. When RA patients were categorized according to the DAS28-ESR score, PON1 concentration and enzymatic activity were lower whereas N-Hcy-protein was higher in those with high disease activity. PON1 activity and Hcy thiolactone were correlated with DAS28-ESR score and myeloperoxidase concentration. In conclusion, RA is associated with deficiency of PON1 activity and increased protein N-homocyseinylation which may contribute to accelerated development of cardiovascular diseases.