Merck
  • Preferential dipeptide incorporation of Porphyromonas gingivalis mediated by proton-dependent oligopeptide transporter (Pot).
Preferential dipeptide incorporation of Porphyromonas gingivalis mediated by proton-dependent oligopeptide transporter (Pot).
FEMS microbiology letters (2020-12-19)
Yuko Ohara-Nemoto, Mohammad Tanvir Sarwar, Yu Shimoyama, Takeshi Kobayakawa, Takayuki K Nemoto
ABSTRACT
Multiple dipeptidyl-peptidases (DPPs) are present in the periplasmic space of Porphyromonas gingivalis, an asaccharolytic periodontopathic bacterium. Dipeptides produced by DPPs are presumed to be transported into the bacterial cells and metabolized to generate energy and cellular components. The present study aimed to identify a transporter responsible for dipeptide uptake in the bacterium. A real-time metabolic analysis demonstrated that P. gingivalis preferentially incorporated Gly-Xaa dipeptides, and then, single amino acids, tripeptides and longer oligopeptides to lesser extents. Heterologous expression of the P. gingivalis serine/threonine transporter (SstT; PGN_1460), oligopeptide transporter (Opt; PGN_1518) and proton-dependent oligopeptide transporter (Pot; PGN_0135) genes demonstrated that Escherichia coli expressing Pot exclusively incorporated Gly-Gly, while SstT managed Ser uptake and Opt was responsible for Gly-Gly-Gly uptake. Dipeptide uptake was significantly decreased in a P. gingivalis Δpot strain and further suppressed in a Δpot-Δopt double-deficient strain. In addition, the growth of the Δpot strain was markedly attenuated and the Δpot-Δopt strain scarcely grew, whereas the ΔsstT strain grew well almost like wild type. Consequently, these results demonstrate that predominant uptake of dipeptide in P. gingivalis is mostly managed by Pot. We thus propose that Pot is a potential therapeutic target of periodontal disease and P. gingivalis-related systemic diseases.
Sigma-Aldrich
Elastin-orcein, elastase substrate
Pricing and availability is not currently available.
Sigma-Aldrich
Gly-Glu
Pricing and availability is not currently available.
Sigma-Aldrich
Glu-Glu
Pricing and availability is not currently available.
Sigma-Aldrich
Gly-Pro
Pricing and availability is not currently available.