Merck
  • Serum proteins in chronic hepatitis B patients treated with peginterferon alfa-2b.
Serum proteins in chronic hepatitis B patients treated with peginterferon alfa-2b.
World journal of gastroenterology (2013-08-22)
Sunida Kuakarn, Poorichaya SomParn, Pisit Tangkijvanich, Varocha Mahachai, Visith Thongboonkerd, Nattiya Hirankarn
ABSTRACT
To study the differential protein profile in serum of hepatitis B patients. Serum samples were obtained from patients with chronic hepatitis B who were receiving peginterferon alfa-2b. The serum samples were subjected to albumin depletion and analyzed by two-dimensional gel electrophoresis (2-DE). Differentially expressed protein spots were identified by electrospray ionization-quadrupole time-of-flight mass spectrometry. Alpha-2-HS-glycoprotein, complement component C3c and CD5 antigen were further analyzed by an enzyme-linked immunosorbent assay and immunonephelometry. Nineteen patients with HBeAg-positive chronic hepatitis B (CHB) were studied. These patients were followed for at least 1 year after treatment and were classified according to their treatment response: responders (n = 9) and non-responders (n = 10). 2-DE and MS/MS analysis were performed to compare the serum proteins before initiating peginterferon alfa-2b. From the quantitative analysis of the 2-D gel, 7 proteins were detected between the two groups at different levels before treatment. Among these potential candidates, serum levels of alpha-2-HS-glycoprotein, complement component C3c and CD5 antigen-like precursor were further analyzed. In the validation phase, 23 subjects, 9 sustained responders and 14 non-responders, were recruited. Interestingly, the levels of alpha-2-HS-glycoprotein and complement component C3c were elevated in the serum of the non-responders compared to the responders. Serum alpha-2-HS-glycoprotein and complement component C3c may be potential serum biomarkers in predicting the treatment response of peginterferon alfa-2b in patients with CHB prior to treatment.