Merck
  • IgE-mediated mast cell activation promotes inflammation and cartilage destruction in osteoarthritis.
IgE-mediated mast cell activation promotes inflammation and cartilage destruction in osteoarthritis.
eLife (2019-05-16)
Qian Wang, Christin M Lepus, Harini Raghu, Laurent L Reber, Mindy M Tsai, Heidi H Wong, Ericka von Kaeppler, Nithya Lingampalli, Michelle S Bloom, Nick Hu, Eileen E Elliott, Francesca Oliviero, Leonardo Punzi, Nicholas J Giori, Stuart B Goodman, Constance R Chu, Jeremy Sokolove, Yoshihiro Fukuoka, Lawrence B Schwartz, Stephen J Galli, William H Robinson
ABSTRACT
Osteoarthritis is characterized by articular cartilage breakdown, and emerging evidence suggests that dysregulated innate immunity is likely involved. Here, we performed proteomic, transcriptomic, and electron microscopic analyses to demonstrate that mast cells are aberrantly activated in human and murine osteoarthritic joint tissues. Using genetic models of mast cell deficiency, we demonstrate that lack of mast cells attenuates osteoarthritis in mice. Using genetic and pharmacologic approaches, we show that the IgE/FcεRI/Syk signaling axis is critical for the development of osteoarthritis. We find that mast cell-derived tryptase induces inflammation, chondrocyte apoptosis, and cartilage breakdown. Our findings demonstrate a central role for IgE-dependent mast cell activation in the pathogenesis of osteoarthritis, suggesting that targeting mast cells could provide therapeutic benefit in human osteoarthritis. This article has been through an editorial process in which the authors decide how to respond to the issues raised during peer review. The Reviewing Editor's assessment is that all the issues have been addressed (see decision letter).
Sigma-Aldrich
Mast Cell Degranulation Assay Kit, The Mast Cell Degranulation Assay Kit provides a quick, efficient & sensitive system for evaluation of tryptase activity in culture supernatants, cell lysates or other tryptase-containing samples.