Merck
  • Reduction of Total Brain and Cerebellum Volumes Associated With Neuronal Autoantibodies in Patients With APECED.
Reduction of Total Brain and Cerebellum Volumes Associated With Neuronal Autoantibodies in Patients With APECED.
The Journal of clinical endocrinology and metabolism (2018-10-20)
Antonella Meloni, Giulia Corda, Luca Saba, Gian-Luca Ferri, Stefano Mariotti, Cristina Cocco
ABSTRACT
In autoimmune polyendocrinopathy-candidiasis-ectodermal dystrophy (APECED), autoantibodies (AutoAbs) labeling brain neurons were reported; conversely, brain MRI alterations associated with these AutoAbs were never reported. To describe brain alterations in APECED and to correlate them with AutoAbs against glutamic acid decarboxylase (GAD), tyrosine hydroxylase (TH), and 5-tryptophan hydroxylase (5-HT) neurons. Fourteen Sardinian patients with APECED and age-matched control subjects were recruited for MRI analysis and blood sampling to detect AutoAbs to GAD, TH, and 5-HT neurons by using rat brain sections. The majority of patients (n = 12) were investigated for AutoAbs a decade earlier, and 7 of 12 were positive for AutoAbs to GAD and TH neurons. Patients with APECED had smaller cerebellum and gray matter volumes, with a ventricular enlargement and a total cerebrospinal fluid (CSF) increase, compared with controls (P < 0.01). In 11 of 14 patients, brain abnormalities were associated with AutoAbs to GAD or TH neurons (titer 1:100 to 15,000) that had persisted for 10 years in 7 of 11 patients. AutoAbs to 5-HT neurons were revealed in all patients with AutoAbs to TH neurons. A decrease in whole brain and cerebellum volumes (P = 0.028) was associated with AutoAbs to GAD neurons, and a CSF increase was associated with AutoAbs to GAD and TH/5-HT neurons (P < 0.05). HLA alleles did not appear to be involved in neuronal autoimmunity. Brain alterations and neuronal AutoAbs were observed in 78.6% of Sardinian patients with APECED, suggesting a brain autoimmune reaction. Prolonged clinical follow-up must be conducted for the possible appearance of clinical neurologic consequences.