Merck
  • Temperature-sensitive recombinant subtilisin protease variants that efficiently degrade molecular biology enzymes.
Temperature-sensitive recombinant subtilisin protease variants that efficiently degrade molecular biology enzymes.
FEMS microbiology letters (2020-10-06)
Vanessa C Thompson, Bailey E McGuire, Mia S Frier, Max S G Legg, Tyler W Dyer, Geoff Gudavicius, Sheila Potter, Francis E Nano
ABSTRACT
We used error-prone PCR to generate mutations in a subtilisin protease-encoding gene, and screened for recombinants that expressed temperature-sensitive (TS) variants. From the dozens of mutations that we detected in the recombinant genes we found that those mutations that affected aspartate-75 had the most profound effect on temperature stability. We thus focused our analysis on two variants of subtilisin C, the more heat-sensitive variant 24 (V24), with amino acid changes D75G, L234M and Q274P; and variant 25 (V25), with a single amino acid change, D75A. For V24 a two log-fold reduction in activity occurs in under 10 min at 50°C. For V25, a two log-fold reduction occurs at 60°C, a temperature that reduces the activity of the wild type enzyme by about 30%. The V24 variant fully inactivates enzymes commonly used in molecular biology research and in molecular diagnostics, and is stabilized against autolysis with propylene glycol concentrations of 10% or greater. The subtilisin variants are produced by a strain of Bacillus subtilis that lacks expression of its native secreted proteases, and the variants can be isolated from the supernatants using nickel affinity chromatography.